missing translation for 'onlineSavingsMsg'
Learn More
Benzoin, 98%, Thermo Scientific Chemicals Product Code.: 10457860

Benzoin, 98%, Thermo Scientific Chemicals

Product Code. 10457860
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10457860

Brand: Thermo Scientific Acros 105511000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004496
InChI Key ISAOCJYIOMOJEB-UHFFFAOYNA-N
Synonym benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketoneShow MoreShow Less
PubChem CID 8400
ChEBI CHEBI:17682
IUPAC Name 2-hydroxy-1,2-diphenylethanone
SMILES OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 133.0°C to 137.0°C
Color White to Yellow
Boiling Point 194.0°C (12.0 mmHg)
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CH(OH)COC6H5
Quantity 100 g
Merck Index 15, 1095
Solubility Information Solubility in water: sparingly soluble
Formula Weight 212.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzoin, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462349 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number105510025,105515000,105511000
Lot NumberA0455778 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number105510025,105515000,105511000
Lot NumberA0398411 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105510025,105515000,105511000
Lot NumberA0393406 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105510025,105515000,105511000
Lot NumberA0405640 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number105510025,105515000,105511000
Product Identifier
  • Benzoin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzoin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.