missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoin methyl ether, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Descripción
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.
Solubility
Soluble in methanol.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.
Solubility
Soluble in methanol.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Especificaciones
Melting Point | 46°C to 50°C |
Density | 1.128 |
Boiling Point | 188°C to 189°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 500 g |
Beilstein | 2052112 |
Merck Index | 14,1093 |
Solubility Information | Soluble in methanol. |
Formula Weight | 226.28 |
Percent Purity | 97% |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'