missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzophenone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 47°C to 50°C |
Density | 1.111 g/mL |
Boiling Point | 305°C |
Flash Point | 138°C (280°F) |
Assay Percent Range | 99% |
Quantity | 250 g |
UN Number | UN3077 |
Beilstein | 1238185 |
Merck Index | 14,1098 |
Solubility Information | Soluble in ethanol and diethyl ether. Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'