Learn More
Benzophenone oxime, 98%, Thermo Scientific Chemicals
£24.80 - £76.90
Chemical Identifiers
CAS | 574-66-3 |
---|---|
Molecular Formula | C13H11NO |
Molecular Weight (g/mol) | 197.24 |
MDL Number | MFCD00051461 |
InChI Key | DNYZBFWKVMKMRM-UHFFFAOYSA-N |
Synonym | benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime |
PubChem CID | 11324 |
IUPAC Name | N-benzhydrylidenehydroxylamine |
SMILES | ON=C(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10707782
|
Thermo Scientific Acros
342330050 |
5 g | Glass bottle |
£24.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10439091
|
Thermo Scientific Acros
342330250 |
25 g | Glass bottle |
£76.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
574-66-3 | |
197.24 | |
DNYZBFWKVMKMRM-UHFFFAOYSA-N | |
11324 | |
ON=C(C1=CC=CC=C1)C1=CC=CC=C1 |
C13H11NO | |
MFCD00051461 | |
benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime | |
N-benzhydrylidenehydroxylamine |
Specifications
574-66-3 | |
100.0 | |
White | |
98% | |
C6H5CNOH(C6H5) | |
15, 1100 | |
Solubility in water: insoluble. Other solubilities: insoluble in hexane,soluble in benzene(warm),chloroform,,dichloromethane,ethyl acetate,methanol | |
ON=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
197.24 | |
197.24 | |
Powder |
97.5 | |
140°C to 144°C | |
Authentic | |
C13H11NO | |
MFCD00051461 | |
benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime | |
DNYZBFWKVMKMRM-UHFFFAOYSA-N | |
N-benzhydrylidenehydroxylamine | |
11324 | |
98% | |
Benzophenone oxime, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-373-6
missing translation for 'rtecsNumber' : DJ1810000
missing translation for 'tsca' : TSCA
- Benzophenone oxime
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.