missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacolone, 97%, Thermo Scientific™

Product Code. 11381377
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 466-37-5
Molecular Formula C26H20O
Molecular Weight (g/mol) 348.445
MDL Number MFCD00004762
InChI Key CFBBKHROQRFCNZ-UHFFFAOYSA-N
Synonym 2,2,2-triphenylacetophenone, tetraphenylethanone, benzopinacolone, ethanone, tetraphenyl, phenyl trityl ketone, acetophenone, 2,2,2-triphenyl, 1,2,2,2-tetraphenylethan-1-one, ss-benzpinakolin, beta-benzopinacolone, .beta.-benzopinacolone
PubChem CID 68049
IUPAC Name 1,2,2,2-tetraphenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 181°C to 182°C
Quantity 5 g
Beilstein 2058376
Formula Weight 348.45
Percent Purity 97%
Chemical Name or Material Benzopinacolone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'