missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoquinonium dibromide, Tocris Bioscience™
Nicotinic antagonist
Chemical Identifiers
CAS | 311-09-1 |
---|---|
Molecular Formula | C34H50Br2N4O2 |
Molecular Weight (g/mol) | 706.608 |
InChI Key | DVQZHWAYOLXTHE-UHFFFAOYSA-N |
Synonym | benzoquinonium dibromide, n,n'-3,6-dioxo-1,4-cyclohexadiene-1,4-diyl bis imino-3,1-propanediyl bis n,n-diethylbenzenemethanaminium dibromide |
PubChem CID | 56972152 |
IUPAC Name | benzyl-[3-[[4-[3-[benzyl(diethyl)azaniumyl]propylamino]-3,6-dioxocyclohexa-1,4-dien-1-yl]amino]propyl]-diethylazanium;dibromide |
SMILES | CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)CC2=CC=CC=C2)CC3=CC=CC=C3.[Br-].[Br-] |
Description
Nicotinic antagonist, ganglion blocker. Activates single channels but blocks open channels at nicotinic receptors.Chemical Identifiers
311-09-1 | |
706.608 | |
benzoquinonium dibromide, n,n'-3,6-dioxo-1,4-cyclohexadiene-1,4-diyl bis imino-3,1-propanediyl bis n,n-diethylbenzenemethanaminium dibromide | |
benzyl-[3-[[4-[3-[benzyl(diethyl)azaniumyl]propylamino]-3,6-dioxocyclohexa-1,4-dien-1-yl]amino]propyl]-diethylazanium;dibromide |
C34H50Br2N4O2 | |
DVQZHWAYOLXTHE-UHFFFAOYSA-N | |
56972152 | |
CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)CC2=CC=CC=C2)CC3=CC=CC=C3.[Br-].[Br-] |
Specifications
311-09-1 | |
benzoquinonium dibromide, n,n'-3,6-dioxo-1,4-cyclohexadiene-1,4-diyl bis imino-3,1-propanediyl bis n,n-diethylbenzenemethanaminium dibromide | |
CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)CC2=CC=CC=C2)CC3=CC=CC=C3.[Br-].[Br-] | |
706.608 | |
706.6 | |
Benzoquinonium dibromide |
C34H50Br2N4O2 | |
DVQZHWAYOLXTHE-UHFFFAOYSA-N | |
benzyl-[3-[[4-[3-[benzyl(diethyl)azaniumyl]propylamino]-3,6-dioxocyclohexa-1,4-dien-1-yl]amino]propyl]-diethylazanium;dibromide | |
56972152 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature