missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl bromide, 97%

Product Code. 10527563 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10527563

Brand: Thermo Scientific Acros 160950250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-32-6 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-32-6
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00000112
InChI Key AQIHMSVIAGNIDM-UHFFFAOYSA-N
Synonym benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide
PubChem CID 12056
IUPAC Name benzoyl bromide
SMILES BrC(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -24.0°C
Color Orange
Density 1.5700g/mL
Boiling Point 218.0°C to 219.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COBr
Refractive Index 1.5880 to 1.5900
Quantity 25 mL
Beilstein 09,195
Specific Gravity 1.57
Solubility Information Solubility in water: may decompose. Other solubilities: miscible in ethyl ether
Formula Weight 185.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzoyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • Benzoyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.