missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetone, 98+%, Thermo Scientific Chemicals

Product Code. 11401908
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 55°C to 60°C
Density 1.09
Boiling Point 260°C to 262°C
Quantity 1000 g
Beilstein 742413
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity ≥98%
Chemical Name or Material Benzoylacetone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'