missing translation for 'onlineSavingsMsg'
Learn More
Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside, Thermo Scientific™ Product Code.: 15440687

Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside, Thermo Scientific™

Product Code. 15440687
500 mg
Click to view available options
Quantity:
500 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15440687

Brand: Thermo Scientific Alfa Aesar J66981.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13343-63-0
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.443
MDL Number MFCD00143679
InChI Key NXGXFAKJUWEFEC-NVZUTRPHSA-N
Synonym n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-7-yl acetamide, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glcopyranoside, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glucopyranoside, n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-7-yl acetamide, maybridge1_006454, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, phenylmethyl 2-acetylamino-2-deoxy-4,6-o-phenylmethylene-, 1-o-benzyl-n-acetyl-4-o,6-o-benzylidene-alpha-d-glucosamine, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-a-d-glcopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-Show MoreShow Less
PubChem CID 2735220
IUPAC Name N-[(4aR,6S,7R,8R,8aS)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
SMILES CC(=O)NC1C(C2C(COC(O2)C3=CC=CC=C3)OC1OCC4=CC=CC=C4)O

Specifications

Melting Point 256°C to 261°C (decomposition)
Color White
Quantity 500 mg
Beilstein 55951
Formula Weight 399.44
Physical Form Crystalline
Chemical Name or Material Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ06G014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberS25E024 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.