missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-aminopiperidine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 15441629
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120278-07-1
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.299
MDL Number MFCD05863884
InChI Key YYIQGSYCCNQAGV-UHFFFAOYSA-N
Synonym 1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775
PubChem CID 17846640
IUPAC Name benzyl 4-aminopiperidine-1-carboxylate
SMILES C1CN(CCC1N)C(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Beilstein 10003356
Solubility Information Slightly soluble in water.
Formula Weight 234.3
Percent Purity 97%
Chemical Name or Material Benzyl 4-aminopiperidine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'