missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 4-aminopiperidine-1-carboxylate, 97%, Thermo Scientific™
£21.40 - £288.00
Chemical Identifiers
CAS | 120278-07-1 |
---|---|
Molecular Formula | C13H18N2O2 |
Molecular Weight (g/mol) | 234.299 |
MDL Number | MFCD05863884 |
InChI Key | YYIQGSYCCNQAGV-UHFFFAOYSA-N |
Synonym | 1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 |
PubChem CID | 17846640 |
IUPAC Name | benzyl 4-aminopiperidine-1-carboxylate |
SMILES | C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461629
|
Thermo Scientific Alfa Aesar
H64424.MD |
250 mg | |||||||
15441629
|
Thermo Scientific Alfa Aesar
H64424.03 |
1 g | |||||||
15451629
|
Thermo Scientific Alfa Aesar
H64424.06 |
5 g |
£288.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
120278-07-1 | |
234.299 | |
YYIQGSYCCNQAGV-UHFFFAOYSA-N | |
17846640 | |
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 |
C13H18N2O2 | |
MFCD05863884 | |
1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 | |
benzyl 4-aminopiperidine-1-carboxylate |
Specifications
120278-07-1 | |
MFCD05863884 | |
1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 | |
YYIQGSYCCNQAGV-UHFFFAOYSA-N | |
benzyl 4-aminopiperidine-1-carboxylate | |
17846640 | |
97% |
C13H18N2O2 | |
10003356 | |
Slightly soluble in water. | |
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 | |
234.299 | |
234.3 | |
Benzyl 4-aminopiperidine-1-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures