missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 4-chlorophenyl ketone, 98%, Thermo Scientific Chemicals
£32.20 - £325.00
Chemical Identifiers
CAS | 1889-71-0 |
---|---|
Molecular Formula | C14H11ClO |
Molecular Weight (g/mol) | 230.69 |
MDL Number | MFCD00016342 |
InChI Key | DXVALSKCLLBZEB-UHFFFAOYSA-N |
Synonym | benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by |
PubChem CID | 233840 |
IUPAC Name | 1-(4-chlorophenyl)-2-phenylethanone |
SMILES | ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488516
|
Thermo Scientific Alfa Aesar
A15959.06 |
5 g |
£32.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498516
|
Thermo Scientific Alfa Aesar
A15959.14 |
25 g |
£108.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408526
|
Thermo Scientific Alfa Aesar
A15959.22 |
100 g |
£325.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1889-71-0 | |
230.69 | |
DXVALSKCLLBZEB-UHFFFAOYSA-N | |
233840 | |
ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 |
C14H11ClO | |
MFCD00016342 | |
benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by | |
1-(4-chlorophenyl)-2-phenylethanone |
Specifications
1889-71-0 | |
C14H11ClO | |
976443 | |
DXVALSKCLLBZEB-UHFFFAOYSA-N | |
1-(4-chlorophenyl)-2-phenylethanone | |
233840 | |
98% |
103°C to 107°C | |
MFCD00016342 | |
benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by | |
ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 | |
230.69 | |
230.69 | |
Benzyl 4-chlorophenyl ketone |
Safety and Handling
P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
H317-H318
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only