missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxytetrahydro-1(2H)-pyridinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10661683
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 95798-23-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
MDL Number MFCD01863722
InChI Key JKIUUDJOCYHIGY-UHFFFAOYSA-N
Synonym benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
PubChem CID 2794620
IUPAC Name benzyl 4-hydroxypiperidine-1-carboxylate
SMILES OC1CCN(CC1)C(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 97.0
Density 1.554g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.28
Percent Purity 97%
Chemical Name or Material Benzyl 4-hydroxytetrahydro-1(2H)-pyridinecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'