missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxytetrahydro-1(2H)-pyridinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10703633
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 95798-23-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
MDL Number MFCD01863722
InChI Key JKIUUDJOCYHIGY-UHFFFAOYSA-N
Synonym benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
PubChem CID 2794620
IUPAC Name benzyl 4-hydroxypiperidine-1-carboxylate
SMILES OC1CCN(CC1)C(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 97.0
Density 1.554g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 235.28
Percent Purity 97%
Chemical Name or Material Benzyl 4-hydroxytetrahydro-1(2H)-pyridinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.