missing translation for 'onlineSavingsMsg'
Learn More
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals Product Code.: 11304589

Benzyl acetoacetate, 97%, Thermo Scientific Chemicals

Product Code. 11304589
25 g
Click to view available options
Quantity:
25 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 11304589

Brand: Thermo Scientific Alfa Aesar A14915.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5396-89-4
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00008785
InChI Key WOFAGNLBCJWEOE-UHFFFAOYSA-N
Synonym benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
PubChem CID 142266
IUPAC Name benzyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Density 1.113
Boiling Point 276°C
Flash Point 108°C (226°F)
Refractive Index 1.512
Quantity 25 g
Beilstein 2049615
Solubility Information It is soluble in alkali solutions.
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material Benzyl acetoacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.