Learn More
Benzyl Alcohol, 99%, Pure, Thermo Scientific Chemicals
£36.40 - £464.00
Chemical Identifiers
CAS | 100-51-6 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00004599,MFCD03792087 |
InChI Key | WVDDGKGOMKODPV-UHFFFAOYSA-N |
Synonym | benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol |
PubChem CID | 244 |
ChEBI | CHEBI:17987 |
IUPAC Name | phenylmethanol |
SMILES | OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011650
|
Thermo Scientific Acros
148390010 |
1 L | Glass bottle |
£36.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10193570
|
Thermo Scientific Acros
148390025 |
2.5 L | Glass bottle |
£58.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10639103
|
Thermo Scientific Acros
148390250 |
25 L | Metal drum |
£464.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-51-6 | |
108.14 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
244 | |
phenylmethanol |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
CHEBI:17987 | |
OCC1=CC=CC=C1 |
Specifications
100-51-6 | |
100.0 | |
Authentic | |
C7H8O | |
1.5380 to 1.5410 (20°C, 589nm) | |
06, 428 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
OCC1=CC=CC=C1 | |
108.14 | |
CHEBI:17987 | |
0.13mbar at 20°C | |
Pure | |
Benzyl alcohol |
98.5 | |
Colorless to Yellow | |
99% | |
C6H5CH2OH | |
MFCD00004599,MFCD03792087 | |
15, 1127 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
108.14 | |
≥98.5% | |
Liquid |
Safety and Handling
GHS P Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Benzyl alcohol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only