missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -16°C to -13°C |
Density | 1.045g/mL at 25°C |
Boiling Point | 203°C to 205°C |
Flash Point | 96°C (204.8°F) |
Ignition Residue | ≤0.005% |
Ignition Point | 817°F |
Packaging | Glass Bottle |
Linear Formula | C6H5CH2OH |
Refractive Index | n20/D 1.539 |
Quantity | 100 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'