missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
Alkalinity | 0.002 meq/g max. |
Acidity | 0.001 meq/g max. |
Melting Point | -15.0°C |
Density | 1.0440g/mL |
Boiling Point | 205.0°C |
Flash Point | 96°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | C6H5CH2OH |
Refractive Index | 1.5380 to 1.5410 |
Show More |
Safety and Handling
Product Identifier
- Benzyl alcohol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction