missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Multi-Compendial, N.F., J.T. Baker™

£170.00 - £1090.00

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15508514
View Documents
Avantor J.T.Baker
9039-10
100 mL Glass Bottle
£170.00
100mL
Please to purchase this item. Need a web account? Register with us today!
15518514
View Documents
Avantor J.T.Baker
1 L Narrow Mouth Amber Glass Bottle, w/ Septum
£828.00
1 L
Please to purchase this item. Need a web account? Register with us today!
15548614
View Documents
Avantor J.T.Baker
4 L Narrow Mouth Amber Glass Bottle
£1090.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

100-51-6
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol
C7H8O
MFCD00004599,MFCD03792087
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
CHEBI:17987
OCC1=CC=CC=C1

Specifications

100-51-6
C7H8O
1L = 1.05kg
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14
Benzyl Alcohol
93.00°C
MFCD00004599,MFCD03792087
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
OCC1=CC=CC=C1
108.14
CHEBI:17987
Multi-Compendial/NF
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Benzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Phenol

SDS
Documents

Documents

Product Certifications