missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 100-51-6 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00004599,MFCD03792087 |
InChI Key | WVDDGKGOMKODPV-UHFFFAOYSA-N |
Synonym | benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol |
PubChem CID | 244 |
ChEBI | CHEBI:17987 |
IUPAC Name | phenylmethanol |
SMILES | OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15602500
|
Honeywell Riedel-de Haën
24122-1L |
1 L |
£85.84
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15612500
|
Honeywell Riedel-de Haën
24122-2.5L |
2.5 L |
£226.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622500
|
Honeywell Riedel-de Haën
24122-30KG |
30 kg |
£802.38
30kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
100-51-6 | |
108.14 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
244 | |
phenylmethanol |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
CHEBI:17987 | |
OCC1=CC=CC=C1 |
Specifications
100-51-6 | |
96°C (204.8°F) | |
817°F | |
C6H5CH2OH | |
3334 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
OCC1=CC=CC=C1 | |
108.14 | |
3.7 (vs air) | |
108.14g/mol | |
99 to 100.5% (GC) | |
Benzyl Alcohol, Meets analytical specification of Ph.Eur.BP, NF |
1.045g/mL at 25°C | |
≤0.005% (as SO4) | |
C7H8O | |
MFCD00004599,MFCD03792087 | |
878307 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
CHEBI:17987 | |
13.3mmHg (100°C), 3.75mmHg (77°C) | |
puriss. |
Safety and Handling
P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338
H302 + H332-H319
Warning
missing translation for 'einecsNumber' : 202-859-9
missing translation for 'rtecsNumber' : DN3150000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Benzyl alcohol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302 + H332-Harmful if swallowed or if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title