missing translation for 'onlineSavingsMsg'
Learn More

Benzyl azide, 94%, Thermo Scientific Chemicals

Product Code. 11392878
2 g
Click to view available options
Quantity:
2 g
10 g
50 g

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-79-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00013836
InChI Key UDLLFLQFQMACJB-UHFFFAOYSA-N
Synonym azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029Show MoreShow Less
PubChem CID 12152
IUPAC Name azidomethylbenzene
SMILES C1=CC=C(C=C1)CN=[N+]=[N-]

Specifications

Density 1.0655
Boiling Point 82°C to 85°C (16 mm)
Flash Point 43°C (110°F)
Assay Percent Range 94%
Refractive Index 1.5341
Quantity 2 g
Solubility Information Insoluble in waterMiscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 133.15
Physical Form Liquid
Chemical Name or Material Benzyl azide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX11K032 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number032635.18,032635.09,032635.04
Lot NumberU23K010 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number032635.18,032635.09,032635.04
Lot NumberN13K047 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number032635.18,032635.09,032635.04
Lot NumberY07J042 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number032635.18,032635.09,032635.04
Lot NumberU29I025 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog Number032635.18,032635.09,032635.04
compliance-icons
Product Identifier
  • Benzyl azide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Benzyl azide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl azide, 94%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.