Learn More
Chemical Identifiers
CAS | 622-79-7 |
---|---|
Molecular Formula | C7H7N3 |
Molecular Weight (g/mol) | 133.154 |
MDL Number | MFCD00013836 |
InChI Key | UDLLFLQFQMACJB-UHFFFAOYSA-N |
Synonym | azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 |
PubChem CID | 12152 |
IUPAC Name | azidomethylbenzene |
SMILES | C1=CC=C(C=C1)CN=[N+]=[N-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392878
|
Thermo Scientific Alfa Aesar
032635.04 |
2 g |
£52.76
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382878
|
Thermo Scientific Alfa Aesar
032635.09 |
10 g |
£186.93
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372878
|
Thermo Scientific Alfa Aesar
032635.18 |
50 g |
£781.89
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
622-79-7 | |
133.154 | |
UDLLFLQFQMACJB-UHFFFAOYSA-N | |
12152 | |
C1=CC=C(C=C1)CN=[N+]=[N-] |
C7H7N3 | |
MFCD00013836 | |
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 | |
azidomethylbenzene |
Specifications
622-79-7 | |
82°C to 85°C (16 mm) | |
94% | |
1.5341 | |
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 | |
UDLLFLQFQMACJB-UHFFFAOYSA-N | |
azidomethylbenzene | |
12152 | |
Liquid |
1.0655 | |
43°C (110°F) | |
C7H7N3 | |
MFCD00013836 | |
Insoluble in waterMiscible with ethanol and diethyl ether. Immiscible with water. | |
C1=CC=C(C=C1)CN=[N+]=[N-] | |
133.154 | |
133.15 | |
Benzyl azide |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'rtecsNumber' : XS6650000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Benzyl azide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : Benzyl azide
RUO – Research Use Only