missing translation for 'onlineSavingsMsg'
Learn More

Benzyl benzoate, 99+%, Thermo Scientific Chemicals

Product Code. 11382076
Click to view available options
Quantity:
250 g
1000 g
2500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Benzyl benzoate is used as repellent for chiggers, ticks, and mosquitoes. It is an effective and inexpensive topical treatment for human scabies. It has vasodilating and spasmolytic effects and is present in many asthma and whooping cough drugs. It is also a dye carrier, solvent for cellulose derivatives, plasticizer, and fixative in the perfume industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with ethanol, alcohol, chloroform, ether, oils.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
Synonym ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
PubChem CID 2345
ChEBI CHEBI:41237
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 18°C to 20°C
Density 1.112
Boiling Point 323°C to 324°C
Flash Point 147°C (296°F)
Linear Formula C6H5CO2CH2C6H5
Refractive Index 1.568
Quantity 1000 g
UN Number UN3082
Beilstein 2049280
Merck Index 14,1127
Solubility Information Miscible with ethanol,alcohol,chloroform,ether,oils.
Formula Weight 212.25
Percent Purity ≥99%
Chemical Name or Material Benzyl benzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'