missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl benzoate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Acidity | 0.3mg KOH/g max. |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | 18.0°C to 20.0°C |
Color | Colorless |
Density | 1.1180g/mL |
Boiling Point | 323.0°C |
Flash Point | 147°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Show More |
Safety and Handling
Product Identifier
- Benzyl benzoate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction