Learn More
Benzyl bromide, 98%, Thermo Scientific Chemicals
£8.70 - £257.28
Chemical Identifiers
CAS | 100-39-0 |
---|---|
MDL Number | MFCD00000172 |
InChI Key | AGEZXYOZHKGVCM-UHFFFAOYSA-N |
Synonym | benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite |
PubChem CID | 7498 |
ChEBI | CHEBI:59858 |
IUPAC Name | bromomethylbenzene |
SMILES | C1=CC=C(C=C1)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10038531
|
Thermo Scientific Acros
105870050 |
5 mL | Glass bottle | N/A | |||||
10418440
|
Thermo Scientific Acros
105871000 |
100 mL | Glass bottle |
£55.78
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366260
|
Thermo Scientific Acros
105875000 |
500 mL | Glass bottle |
£154.77
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10767583
|
Thermo Scientific Acros
105870010 |
1 L | Glass bottle |
£257.28
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
100-39-0 | |
AGEZXYOZHKGVCM-UHFFFAOYSA-N | |
7498 | |
bromomethylbenzene |
MFCD00000172 | |
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite | |
CHEBI:59858 | |
C1=CC=C(C=C1)CBr |
Specifications
100-39-0 | |
100.0 | |
Authentic | |
C7H7Br | |
1.5745 to 1.5765 (20°C, 589nm) | |
05, 306 | |
15, 1131 | |
AGEZXYOZHKGVCM-UHFFFAOYSA-N | |
bromomethylbenzene | |
7498 | |
171.04 | |
Liquid |
97.5 | |
Colorless to Yellow | |
98% | |
C6H5CH2Br | |
MFCD00000172 | |
05,25; 13,51 | |
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite | |
C1=CC=C(C=C1)CBr | |
171.037 | |
CHEBI:59858 | |
≥97.5% | |
Benzyl bromide, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-847-3
missing translation for 'rtecsNumber' : XS7965000
missing translation for 'tsca' : TSCA
RUO – Research Use Only