missing translation for 'onlineSavingsMsg'
Learn More

Benzyl bromide, 99%, Thermo Scientific Chemicals

£20.80 - £153.77

Chemical Identifiers

CAS 100-39-0
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.037
MDL Number MFCD00000172
InChI Key AGEZXYOZHKGVCM-UHFFFAOYSA-N
Synonym benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
PubChem CID 7498
ChEBI CHEBI:59858
IUPAC Name bromomethylbenzene
SMILES C1=CC=C(C=C1)CBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405653
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13535.18
50 g
£20.80
50g
Please to purchase this item. Need a web account? Register with us today!
11415653
View Documents
Thermo Scientific Alfa Aesar
A13535.30
250 g
£47.44
250g
Please to purchase this item. Need a web account? Register with us today!
11495643
View Documents
Thermo Scientific Alfa Aesar
A13535.0B
1000 g
£153.77
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

Solubility
Miscible with benzene, carbon tetrachloride, ethanol and ether.

Notes
Light and moisture sensitive. Incompatible with bases, alcohols, amines, oxidizing agents and corrodes steel.
Specifications

Chemical Identifiers

100-39-0
171.037
AGEZXYOZHKGVCM-UHFFFAOYSA-N
7498
bromomethylbenzene
C7H7Br
MFCD00000172
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
CHEBI:59858
C1=CC=C(C=C1)CBr

Specifications

100-39-0
1.438 g/mL
87°C (189°F)
C7H7Br
MFCD00000172
Moisture sensitive
Miscible with benzene,carbon tetrachloride,ethanol and ether.
C1=CC=C(C=C1)CBr
171.037
CHEBI:59858
Liquid
-4°C
198°C to 199°C
99%
1.575
385801
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
AGEZXYOZHKGVCM-UHFFFAOYSA-N
bromomethylbenzene
7498
171.04
Benzyl Bromide
Videos
Safety and Handling

Safety and Handling

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 202-847-3

missing translation for 'rtecsNumber' : XS7965000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Moisture sensitive

Recommended Storage : Ambient temperatures

Product Identifier
  • Benzyl bromide

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.