Learn More
Benzyl chloride, 99%, pure, stabilized, Thermo Scientific Chemicals
£16.70 - £268.50
Chemical Identifiers
CAS | 100-44-7, 75-56-9 |
---|---|
MDL Number | MFCD00000889 |
InChI Key | KCXMKQUNVWSEMD-UHFFFAOYSA-N |
Synonym | benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol |
PubChem CID | 7503 |
ChEBI | CHEBI:615597 |
IUPAC Name | chloromethylbenzene |
SMILES | C1=CC=C(C=C1)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10592052
|
Thermo Scientific Acros
405090050 |
5 g | Glass Bottle |
£16.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10720482
|
Thermo Scientific Acros
405092500 |
250 g | Glass Bottle |
£16.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10065740
|
Thermo Scientific Acros
405090010 |
1 kg | Glass Bottle |
£36.10
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10167323
|
Thermo Scientific Acros
405090025 |
2.5 kg | Glass Bottle |
£73.20
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15179283
|
Thermo Scientific Acros
405090100 |
10 kg | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
100-44-7, 75-56-9 | |
KCXMKQUNVWSEMD-UHFFFAOYSA-N | |
7503 | |
chloromethylbenzene |
MFCD00000889 | |
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol | |
CHEBI:615597 | |
C1=CC=C(C=C1)CCl |
Specifications
100-44-7, 75-56-9 | |
100.0 | |
Authentic | |
C7H7Cl | |
1.5370 to 1.5390 (20°C, 589nm) | |
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol | |
C1=CC=C(C=C1)CCl | |
0.25% propylene oxide | |
7503 | |
126.59 | |
Pure | |
Benzyl chloride, Stabilized, Pure |
98.5 | |
Colorless to Yellow | |
99% | |
C6H5CH2Cl | |
MFCD00000889 | |
KCXMKQUNVWSEMD-UHFFFAOYSA-N | |
chloromethylbenzene | |
126.583 | |
CHEBI:615597 | |
≥98.5% | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Benzyl chloride, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only