Learn More
Benzyl chloromethyl ether, 75%, Thermo Scientific Chemicals
£28.20 - £333.00
Chemical Identifiers
CAS | 3587-60-8 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.61 |
MDL Number | MFCD00000886 |
InChI Key | LADPCMZCENPFGV-UHFFFAOYSA-N |
Synonym | benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl |
PubChem CID | 137983 |
IUPAC Name | chloromethoxymethylbenzene |
SMILES | C1=CC=C(C=C1)COCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10083892
|
Thermo Scientific Acros
376950100 |
10 g | Glass bottle |
£28.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310933
|
Thermo Scientific Acros
376950500 |
50 g | Glass bottle |
£85.80
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247852
|
Thermo Scientific Acros
376952500 |
250 g | Glass bottle |
£333.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3587-60-8 | |
156.61 | |
LADPCMZCENPFGV-UHFFFAOYSA-N | |
137983 | |
C1=CC=C(C=C1)COCCl |
C8H9ClO | |
MFCD00000886 | |
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl | |
chloromethoxymethylbenzene |
Specifications
3587-60-8 | |
100.0 | |
1.1300g/mL | |
91°C | |
70% min. (GC) | |
C6H5CH2OCH2Cl | |
1.13 | |
LADPCMZCENPFGV-UHFFFAOYSA-N | |
chloromethoxymethylbenzene | |
137983 | |
75% | |
Benzyl chloromethyl ether |
70.0 | |
Colorless to Yellow | |
102°C (14.0 mmHg) | |
Authentic | |
C8H9ClO | |
MFCD00000886 | |
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl | |
C1=CC=C(C=C1)COCCl | |
156.61 | |
156.61 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause cancer.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handl
GHS Signal Word: Danger
- Benzyl chloromethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only