missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ether, 99%

Product Code. 10113630 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10113630

Brand: Thermo Scientific Acros 148400010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-50-4 | C14H14O | 198.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-50-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.26
MDL Number MFCD00004780
InChI Key MHDVGSVTJDSBDK-UHFFFAOYSA-N
Synonym dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
PubChem CID 7657
ChEBI CHEBI:87411
IUPAC Name phenylmethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.5°C to 3.5°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 298.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5CH2)2O
Refractive Index 1.5610 to 1.5630
Quantity 1 L
Beilstein 06, 434
Merck Index 15, 1135
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and chloroform
Formula Weight 198.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl ether, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl ether
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.