missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ethyl malonate, tech. 85%, Thermo Scientific Chemicals

Product Code. 11446186
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzyl ethyl malonate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl ethyl malonate is used as a pharmaceutical intermediate.

Solubility
Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42998-51-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009194
InChI Key CGNOCUSLPSCMLL-UHFFFAOYSA-N
Synonym benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate
PubChem CID 562228
SMILES CCOC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Boiling Point 145°C (4 mmHg)
Refractive Index 1.497
Quantity 5 g
Solubility Information Soluble in water 525.7mg/L @ 25°C. Soluble in alcohol.
IUPAC Name 3-O-benzyl 1-O-ethyl propanedioate
Formula Weight 222.24
Percent Purity 85%
Grade Technical
Chemical Name or Material Benzyl ethyl malonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'