missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isocyanate, 98%

Product Code. 11327775
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11327775

Brand: Thermo Scientific Alfa Aesar L02633.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3173-56-6 | C8H7NO | 133.15 g/mol

Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

Solubility
It hydrolyzes with water.

Notes
Moisture& heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3173-56-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00009701
InChI Key YDNLNVZZTACNJX-UHFFFAOYSA-N
Synonym benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
PubChem CID 76639
IUPAC Name isocyanatomethylbenzene
SMILES O=C=NCC1=CC=CC=C1

Specifications

Density 1.078
Boiling Point 53°C to 54°C (0.8 mmHg)
Flash Point 43°C (109°F)
Odor Pungent
Linear Formula C6H5CH2NCO
Refractive Index 1.525
Quantity 5 g
UN Number UN3080
Beilstein 636508
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material Benzyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.