Learn More
Benzyl isocyanate, 98%, Thermo Scientific Chemicals
£20.10 - £147.00
Chemical Identifiers
CAS | 3173-56-6 |
---|---|
Molecular Formula | C8H7NO |
Molecular Weight (g/mol) | 133.15 |
MDL Number | MFCD00009701 |
InChI Key | YDNLNVZZTACNJX-UHFFFAOYSA-N |
Synonym | benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate |
PubChem CID | 76639 |
IUPAC Name | isocyanatomethylbenzene |
SMILES | O=C=NCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317775
|
Thermo Scientific Alfa Aesar
L02633.03 |
1 g |
£20.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327775
|
Thermo Scientific Alfa Aesar
L02633.06 |
5 g |
£46.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337775
|
Thermo Scientific Alfa Aesar
L02633.14 |
25 g |
£147.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.
Solubility
It hydrolyzes with water.
Notes
Moisture & heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from alcohols, amines.
Chemical Identifiers
3173-56-6 | |
133.15 | |
YDNLNVZZTACNJX-UHFFFAOYSA-N | |
76639 | |
O=C=NCC1=CC=CC=C1 |
C8H7NO | |
MFCD00009701 | |
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate | |
isocyanatomethylbenzene |
Specifications
3173-56-6 | |
53°C to 54°C (0.8 mmHg) | |
Pungent | |
C6H5CH2NCO | |
MFCD00009701 | |
636508 | |
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate | |
YDNLNVZZTACNJX-UHFFFAOYSA-N | |
isocyanatomethylbenzene | |
76639 | |
98% |
1.078 | |
43°C (109°F) | |
C8H7NO | |
1.525 | |
UN3080 | |
Moisture sensitive | |
It hydrolyzes with water. | |
O=C=NCC1=CC=CC=C1 | |
133.15 | |
133.15 | |
Benzyl isocyanate |
Safety and Handling
GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H315-H317-H319-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 221-640-9
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only