Learn More
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Description
Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoylacetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoylacetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Density | 1.125 |
Boiling Point | 242°C to 243°C |
Flash Point | >110°C (230°F) |
Odor | Pungent |
Refractive Index | 1.603 |
Quantity | 5 g |
UN Number | UN2922 |
Beilstein | 386135 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.