Learn More
Benzyl mercaptan, 99%, Thermo Scientific Chemicals
£41.70 - £172.00
Chemical Identifiers
CAS | 100-53-8 |
---|---|
Molecular Formula | C7H8S |
Molecular Weight (g/mol) | 124.20 |
MDL Number | MFCD00004867,MFCD00801588,MFCD01863867 |
InChI Key | UENWRTRMUIOCKN-UHFFFAOYSA-N |
Synonym | benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan |
PubChem CID | 7509 |
IUPAC Name | phenylmethanethiol |
SMILES | SCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10239370
|
Thermo Scientific Acros
421261000 |
100 g | Glass bottle |
£41.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10095980
|
Thermo Scientific Acros
421265000 |
500 g | Glass bottle |
£172.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-53-8 | |
124.20 | |
UENWRTRMUIOCKN-UHFFFAOYSA-N | |
7509 | |
SCC1=CC=CC=C1 |
C7H8S | |
MFCD00004867,MFCD00801588,MFCD01863867 | |
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan | |
phenylmethanethiol |
Specifications
100-53-8 | |
100.0 | |
Colorless to Yellow | |
195°C | |
Authentic | |
C7H8S | |
1.5735 to 1.5775 | |
06, IV, 2632 | |
1.058 | |
Solubility in water: insoluble | |
SCC1=CC=CC=C1 | |
124.20 | |
124.21 | |
Liquid |
98.5 | |
-29°C | |
1.0580g/mL | |
70°C | |
98.5% min. (GC) | |
C6H5CH2SH | |
MFCD00004867,MFCD00801588,MFCD01863867 | |
15, 9475 | |
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan | |
UENWRTRMUIOCKN-UHFFFAOYSA-N | |
phenylmethanethiol | |
7509 | |
99% | |
Benzyl mercaptan |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NO
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-862-5
Recommended Storage : Product darkens during storage
- Benzyl mercaptan
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P264-Wash thoroughly after handling.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only