Learn More
Benzyl phenyl ether, 97%, Thermo Scientific Chemicals
£29.60 - £267.00
Chemical Identifiers
CAS | 946-80-5 |
---|---|
Molecular Formula | C13H12O |
Molecular Weight (g/mol) | 184.238 |
MDL Number | MFCD00020660 |
InChI Key | BOTNYLSAWDQNEX-UHFFFAOYSA-N |
Synonym | benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole |
PubChem CID | 70352 |
IUPAC Name | phenoxymethylbenzene |
SMILES | C1=CC=C(C=C1)COC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415107
|
Thermo Scientific Alfa Aesar
B22539.06 |
5 g |
£29.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425107
|
Thermo Scientific Alfa Aesar
B22539.14 |
25 g |
£88.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435107
|
Thermo Scientific Alfa Aesar
B22539.22 |
100 g |
£267.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
946-80-5 | |
184.238 | |
BOTNYLSAWDQNEX-UHFFFAOYSA-N | |
70352 | |
C1=CC=C(C=C1)COC2=CC=CC=C2 |
C13H12O | |
MFCD00020660 | |
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole | |
phenoxymethylbenzene |
Specifications
946-80-5 | |
286°C to 287°C | |
C13H12O | |
2045713 | |
Insoluble in water. | |
C1=CC=C(C=C1)COC2=CC=CC=C2 | |
184.238 | |
184.24 | |
Benzyl phenyl ether |
37°C to 40°C | |
>110°C (230°F) | |
MFCD00020660 | |
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole | |
BOTNYLSAWDQNEX-UHFFFAOYSA-N | |
phenoxymethylbenzene | |
70352 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only