missing translation for 'onlineSavingsMsg'
Learn More
Benzyltributylammonium chloride, 98+%, Thermo Scientific Chemicals Product Code.: 10164440

Benzyltributylammonium chloride, 98+%, Thermo Scientific Chemicals

Product Code. 10164440
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10164440

Brand: Thermo Scientific Acros 186151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23616-79-7
Molecular Formula C19H34ClN
Molecular Weight (g/mol) 311.94
MDL Number MFCD00011849
InChI Key VJGNLOIQCWLBJR-UHFFFAOYSA-M
Synonym benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbacShow MoreShow Less
PubChem CID 159952
IUPAC Name benzyl(tributyl)azanium;chloride
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 155°C to 163°C
Color White to Ivory
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CH2N[(CH2)3CH3]3Cl
Quantity 100 g
Fieser 06,41
Solubility Information Solubility in water: freely soluble in water
Formula Weight 311.94
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltributylammonium chloride, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463050 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number186155000,186151000
Lot NumberA0444312 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number186155000,186151000
Lot NumberA0456087 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog Number186155000,186151000
Lot NumberA0418305 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number186155000,186151000
Lot NumberA0433841 Certificate TypeCertificate of Analysis Date22/09/2021 Catalog Number
compliance-icons
Product Identifier
  • Benzyltributylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyltributylammonium chloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.