missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltriethylammonium bromide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼200°C (decomposition) |
Quantity | 25 g |
Beilstein | 3739113 |
Sensitivity | Hygroscopic |
Formula Weight | 272.24 |
Percent Purity | ≥98% |
Chemical Name or Material | Benzyltriethylammonium bromide |
Safety and Handling
Product Identifier
- Benzyl triethylammonium bromide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Benzyltriethylammonium bromide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'