missing translation for 'onlineSavingsMsg'
Learn More
Benzyltriethylammonium chloride, 99%, Thermo Scientific Chemicals Product Code.: 11480813

Benzyltriethylammonium chloride, 99%, Thermo Scientific Chemicals

Product Code. 11480813
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11480813

Brand: Thermo Scientific Alfa Aesar A13268.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzyltriethylammonium chloride is used as a catalyst in the preparation of 2-phenylbutyronitrile from phenyl acetonitrile. It is involved in the Knoevenagel condensation of carbonyl compounds with active methylene compounds to give olefinic products. It acts as a phase transfer catalyst used in the alkylation reaction. It reacts with 1H-Pyridine-2-thione to get 2-Benzylsulfanyl-pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriethylammonium chloride is used as a catalyst in the preparation of 2-phenylbutyronitrile from phenyl acetonitrile. It is involved in the Knoevenagel condensation of carbonyl compounds with active methylene compounds to give olefinic products. It acts as a phase transfer catalyst used in the alkylation reaction. It reacts with 1H-Pyridine-2-thione to get 2-Benzylsulfanyl-pyridine.

Solubility
Soluble in acetone, alcohol and water. Insoluble in ether.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-37-1
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.78
MDL Number MFCD00011824
InChI Key HTZCNXWZYVXIMZ-UHFFFAOYSA-M
Synonym benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chlorideShow MoreShow Less
PubChem CID 66133
IUPAC Name benzyl(triethyl)azanium;chloride
SMILES [Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point ∼190°C (decomposition)
Quantity 100 g
Beilstein 3574984
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,alcohol and water. Insoluble in ether.
Formula Weight 227.78
Percent Purity 99%
Chemical Name or Material Benzyltriethylammonium chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251753 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13268.22,A13268.36
Lot Number10250449 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberA13268.22,A13268.36
Lot NumberN24J050 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA13268.22
Lot NumberT22I015 Certificate TypeCertificate of Analysis Date01/08/2022 Catalog NumberA13268.22,A13268.36
Lot Number10240285 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog NumberA13268.22,A13268.36
compliance-icons
Product Identifier
  • Benzyl triethylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Benzyl triethylammonium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyltriethylammonium chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.