Learn More
Benzyltrimethylammonium chloride, 50 wt.% solution in water, Thermo Scientific Chemicals
£12.50 - £40.10
Chemical Identifiers
CAS | 56-93-9, 7732-18-5 |
---|---|
Molecular Formula | C10H16ClN |
Molecular Weight (g/mol) | 185.70 |
MDL Number | MFCD00011782 |
InChI Key | KXHPPCXNWTUNSB-UHFFFAOYSA-M |
Synonym | benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride |
PubChem CID | 5963 |
IUPAC Name | benzyl(trimethyl)azanium;chloride |
SMILES | [Cl-].C[N+](C)(C)CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10430871
|
Thermo Scientific Acros
209381000 |
100 g | Glass bottle |
£12.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229683
|
Thermo Scientific Acros
209380010 |
1 kg | Glass bottle |
£40.10
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
56-93-9, 7732-18-5 | |
185.70 | |
KXHPPCXNWTUNSB-UHFFFAOYSA-M | |
5963 | |
[Cl-].C[N+](C)(C)CC1=CC=CC=C1 |
C10H16ClN | |
MFCD00011782 | |
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride | |
benzyl(trimethyl)azanium;chloride |
Specifications
Colorless to Yellow | |
49.0 | |
C10H16ClN | |
MFCD00011782 | |
01,53 | |
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride | |
[Cl-].C[N+](C)(C)CC1=CC=CC=C1 | |
185.70 | |
185.7 | |
1.445 | |
Liquid |
56-93-9, 7732-18-5 | |
51.0 | |
C6H5CH2N(CH3)3Cl | |
12, 1020 | |
1.0440g/mL | |
KXHPPCXNWTUNSB-UHFFFAOYSA-M | |
benzyl(trimethyl)azanium;chloride | |
5963 | |
≥49% | |
1.044 | |
Benzyltrimethylammonium chloride, 50 wt. % Solution in Water |
Safety and Handling
GHS P Statement Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF exposed: Call a POISON CENTRE or doctor/physician. Avoid release to the environment.
Danger
missing translation for 'einecsNumber' : 200-300-3
missing translation for 'rtecsNumber' : BO8400000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.