missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium dichloroiodate, 99%, Thermo Scientific Chemicals

Product Code. 10130362
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.04
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 128°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2N+(CH3)3Cl2I-
Quantity 5 g
Fieser 15,25; 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methylene chloride
Formula Weight 348.04
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Benzyltrimethylammonium dichloroiodate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Benzyltrimethylammonium dichloroiodate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'