missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltrimethylammonium hydroxide, 20% w/w aq. soln., Thermo Scientific™
Click to view available options
Quantity:
250 mL
1 L
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless to Yellow |
Odor | Amine-like |
Linear Formula | C6H5CH2N(CH3)3OH |
Quantity | 1 L |
UN Number | UN3267 |
Sensitivity | Air Sensitive |
Formula Weight | 167.25 |
Concentration or Composition (by Analyte or Components) | 20% w/w aq. soln. |
Physical Form | Liquid |
Chemical Name or Material | Benzyltrimethylammonium hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'