missing translation for 'onlineSavingsMsg'
Learn More

Bepridil hydrochloride, Tocris Bioscience™

Product Code. 18045922
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective Ca2+ channel blocker; also NCX inhibitor

Calcium channel blocker. Also inhibits Na+/Ca2+ exchange (NCX), sodium channels and cardiac sarcolemmal KATP channels; opens mitochondrial KATP channels. Antiarrhythmic and antihypertensive; acts as a calcium sensitizer.

Specifications

CAS 68099-86-5
Molecular Formula C24H35ClN2O
Quantity 50 mg
Synonym bepridil hydrochloride, cordium, bepridil hcl, 1-isobutoxy-2-pyrrolidino-3-n-benzylanilino propane hydrochloride, 1-pyrrolidineethanamine, beta-2-methylpropoxy methyl-n-phenyl-n-phenylmethyl-, monohydrochloride, bepridil monohydrochloride, n-benzyl-n-3-isobutoxy-2-pyrrolidin-1-yl-propyl aniline hydrochloride, n-benzyl-beta-2-methylpropoxy methyl-n-phenylpyrrolidine-1-ethylamine monohydrochloride, prestwick_697, opera_id_594
InChI Key JXBBWYGMTNAYNM-UHFFFAOYSA-N
SMILES CC(C)COCC(CN(CC1=CC=CC=C1)C2=CC=CC=C2)N3CCCC3.Cl
IUPAC Name N-benzyl-N-[3-(2-methylpropoxy)-2-pyrrolidin-1-ylpropyl]aniline;hydrochloride
Molecular Weight (g/mol) 403.007
PubChem CID 50088
ChEBI CHEBI:52712
Formula Weight 403
Percent Purity >99%
Chemical Name or Material Bepridil hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'