missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Berilium solution for ICP 10g/L HCl matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£1380.00 - £5020.00
Specifications
Concentration | 10±0.2g/L |
---|---|
Type | Mass Spectrometry Standard |
Analyte or Component Names | Be |
Solvent or Matrix | HCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12916264
|
Fisher Chemical
|
100 mL | Bottle |
£1380.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12926264
|
Fisher Chemical
|
250 mL | Bottle |
£2680.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12936264
|
Fisher Chemical
|
500 mL | Bottle |
£5020.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10±0.2g/L | |
Mass Spectrometry Standard | |
Be | |
HCl |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 Months. (12 Months after opening).
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Beryllium (Be) 10000mg/l in HCl 5%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title