missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Berilium solution for ICP 1g/L HCl matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£139.00 - £364.00
Specifications
Concentration | 10±0.2g/L |
---|---|
Type | Mass Spectrometry Standard |
Analyte or Component Names | Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P,Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn |
Solvent or Matrix | HNO3/ HF |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12983364
|
Fisher Chemical
S 007 100 |
100 mL | Bottle |
£139.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12993364
|
Fisher Chemical
S 007 250 |
250 mL | Bottle |
£217.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12903374
|
Fisher Chemical
S 007 500 |
500 mL | Bottle |
£364.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10±0.2g/L | |
Mass Spectrometry Standard | |
Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P,Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn | |
HNO3/ HF |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 Months. (12 Months after opening).
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Beryllium (Be) 1000mg/l in HCl 2%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
Hazard Statement
- H332-Harmful if inhaled.
- H350-May cause cancer.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .