missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Knitted Polyester Lab-Tips Swabs
BIOPHARMA

For cleaning grooved, slotted and recessed areas

Brand:  Berkshire™ LTP125.5

 View more versions of this product

Product Code. 12896804

  • £348.00 / Pack of 500
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Continuous filament polyester knit head
  • Short polypropylene handle with a rigid head
  • Thermally bonded head
  • Laundered and packaged in Class 10 (ISO 4) cleanroom
  • Autoclavable

Specifications

Specifications

ISO Class 4
5 in.
Green
5.5mm
3.94 in.
4.8mm
2.5mm
0.27 in.
Polyester
0.21 in.
Cleaning of sensitive equipment such as print heads, sampling surfaces for cleaning validation
Swab
125mm
100mm
0.98 in.
3.8mm
Rigid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers