missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ SatPax™ 550 Cleanroom Presaturated Wipers

Pre-wetted format provides an easy to use solution

Brand:  Berkshire™ SPX550.001.24

Code : NEW

Additional Details : CAS Number : 67-63-0 Weight : 13.00000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II

Product Code. 15217198

  • £680.00 / Pack of 1200
Estimated Shipment: 11-10-2024
to see stock.

Description

Description

 

  • Removes residue after detergent/disinfectant use
  • Soft, 100% meltblown polypropylene
  • Traceability and process Accountability via lot number
  • Exceptionally low levels of fiber, particles and ions
  • L x W: 11 x 9 in. (27 x 23cm)

 

Specifications

Specifications

SatPax™ 550 Pre-saturated Wipes
27 cm
23 cm
11 x 9 in. (28 x 23 cm)
Isopropyl Alcohol and Deionized Water
11 in.
9 in.
Polypropylene
Disinfectant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers