missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beryllium plasma standard solution, Thermo Scientific Chemicals, Specpure™
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 100 mL |
Color | Colorless |
Molecular Formula | Be4O(C2H3O2)6 in 5% HNO3 |
Concentration or Composition (by Analyte or Components) | Nitric acid: 5.0%; Beryllium oxide acetate: 1.13%; Water: 93.87% |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Physical Form | Liquid |
Chemical Name or Material | Beryllium |
Safety and Handling
Product Identifier
- Beryllium, plasma standard solution, Specpure, Be 1000µg/ml
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H350i-May cause cancer by inhalation.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'