missing translation for 'onlineSavingsMsg'
Learn More
Beryllium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™ Product Code.: 12957253
compliance-icons compliance-icons

Beryllium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™

Product Code. 12957253
100 mL
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy

Product Code. 12957253

Brand: Fisher Chemical S 407 100

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Bottle
Quantity 100 mL
Concentration 100 mg/L
Solvent or Matrix HNO3
Chemical Name or Material Beryllium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1033377 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 407 100
compliance-icons compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. (ICP & ICP-MS). Beryllium (Be) 100mg/l in HNO3 2%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Beryllium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.