missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, 99.8% (metals basis), Thermo Scientific™

Product Code. 15436375
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-41-7
Molecular Formula Be
MDL Number MFCD00134032
Quantity 25 x 25 mm
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
SMILES [Be]
IUPAC Name beryllium
Molecular Weight (g/mol) 9.01
PubChem CID 5460467
ChEBI CHEBI:33783
Percent Purity 99.8%
Odor Odorless
Weight ≈0.012g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, Not certified pinhole free
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, 99.8% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.