missing translation for 'onlineSavingsMsg'
Learn More
Beryllium Ultrathin foil, 1 micron thick, 99.9% (metals basis), Thermo Scientific™ Product Code.: 15436345

Beryllium Ultrathin foil, 1 micron thick, 99.9% (metals basis), Thermo Scientific™

Product Code. 15436345
12.5 x 12.5 mm
Click to view available options
Quantity:
12.5 x 12.5 mm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15436345

Brand: Thermo Scientific Alfa Aesar 040709.0T

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Alloy source, X-ray tube manufacture, neutron source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-41-7
Molecular Formula Be
MDL Number MFCD00134032
Quantity 12.5 x 12.5 mm
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
SMILES [Be]
IUPAC Name beryllium
Molecular Weight (g/mol) 9.01
PubChem CID 5460467
ChEBI CHEBI:33783
Percent Purity 99.9%
Odor Odorless
Particle Size 1 Micron thick
Assay (metals basis)
Chemical Name or Material Beryllium Ultrathin foil
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Beryllium Ultrathin foil, 1 micron thick, 99.9% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.